Would you like to log in again?
Streptomyces hyalurolyticus Hyaluronidase
Supplier: MP Biomedicals
08320421
IC08320421EA 755 USD InStock
IC08320421
Streptomyces hyalurolyticus Hyaluronidase
Enzymes
  • Lyophilized, salt-free
  • Activity: ∼2000 units/mg
  • Protease and lysozyme free
  • Each vial contains 100 U
Enzyme activity is determined by a change in optical density in a hyaluronic acid solution. It is compared to a hyaluronidase International Standard expressed as I.U.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty