Would you like to log in again?
Potato Dextrose Agar, MP Biomedical, LLC.
Supplier: MP Biomedicals
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
091008517
IC091008517EA 190.92 USD InStock
IC091008517
Potato Dextrose Agar, MP Biomedical, LLC.
Media for Microbiology Dehydrated Media for Microbiology
Potato Dextrose Agar is suited for the cultivation of yeasts and molds from dairy products and other foods. It may also be used to induce sporulation in many fungi.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty