Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hanks' Balanced Salts, without Sodium Bicarbonate, MP Biomedical, LLC

Supplier: MP Biomedicals
091710122 091710120
IC091710122EA 48.75 USD InStock
IC091710122 IC091710120
Hanks' Balanced Salts, without Sodium Bicarbonate, MP Biomedical, LLC
Buffers Cell Culture Buffers
Hanks' Balanced Salts solution is used to maintain the pH and osmotic balance in the medium and to provide the cells with water and essential inorganic ions.

  • pH maintenance in culture medium

Soluble in water (9.8 g/L).

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty