Would you like to log in again?
1X Phosphate Buffered Saline without Magnesium and Calcium, MP Biomedicals
Supplier: MP Biomedicals
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
091860454 091860454X6 091860449
IC091860449EA 20.8 USD InStock
IC091860449 IC091860454 IC091860454X6
1X Phosphate Buffered Saline without Magnesium and Calcium, MP Biomedicals
Buffers Cell Culture Buffers
Phosphate Buffered Salts, Dulbecco's Formula (DPBS) is a balanced salt solution (BSS) used for the handling and culturing of mammalian cells.
  • Balanced salt solution
  • Irrigate, wash, and dilute mammalian cells
  • pH maintained in the physiological range
Phosphate Buffered Salts, Dulbecco's Formula (DPBS) is a balanced salt solution (BSS) used for the handling and culturing of mammalian cells. DPBS is used to to irrigate, wash, and dilute mammalian cells. Phosphate buffering maintains the pH in the physiological range. Calcium and magnesium facilitate cell binding and clumping. DPBS without these ions can be used to wash and rinse suspended cells.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty