Would you like to log in again?
Human Type AB Serum (HCV, HIV, HBsAG Free), MP Biomedical
Supplier: MP Biomedicals
092931949
IC092931949EA 345.17 USD InStock
IC092931949
Human Type AB Serum (HCV, HIV, HBsAG Free), MP Biomedical
Sera (Serum)
Human Type AB Serum is used as a growth supplement for cell culture media and in other research applications.
  • Collected from Type AB human blood
  • Liquid media
  • Store at —20 °C
It has been proven to grow many human cell lines at a faster rate and with a smaller percentage of serum than mixed blood group serum.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty