Would you like to log in again?
Catalase
Supplier: MP Biomedicals
0210042801 0210042805
IC10042805EA 1177.37 USD InStock
IC10042805 IC10042801
Catalase
Enzymes
Dissolves readily at 5 mg/mL in 0.05m sodium/potassium phosphate pH 7.0 giving a clear green solution.
One unit decomposes one µmole of H2O2 per minute at 25 °C, pH 7.0.
The catalase test is one of test used by microbiologists to identify species of bacteria.Catalase is also used in the food industry for removing hydrogen peroxide from milk prior to cheese production.
Activates the decomposition of hydrogen peroxide, Natural antioxidant used to study roles of reactive oxygen species in gene expression and apoptosis.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty