Would you like to log in again?
Alpha-Amylase (from Bacillus subtilis)
Supplier: MP Biomedicals
0210044790 0210044725 0210044780
IC10044780EA 159.16 USD InStock
IC10044780 IC10044790 IC10044725
Alpha-Amylase (from Bacillus subtilis)
Enzymes
This α-amylase product is a liquefying diastase supplied as a dry, free-flowing powder having high uniformity and excellent storage stability.
  • Presentation: Tan Powder
  • pH: 5 to 7 (optimum around 6.1)
The general procedure for the enzymatic liquefaction of starch is the same, regardless of the nature of the end product. The degree of conversion is followed by measuring the viscosity. The ultimate viscosity is controlled by the level of enzyme and the time which elapses after gelatinization. The usual conditions are 0.03 - 0.06% a-Amylase based on weight of starch for 15 to 30 minutes. The starch is suspended in cold water with agitation sufficient to assure uniform dispersion. If vigorous agitation is provided, the dry solid enzyme may be added directly to the suspension. A safer practice, however, is to dissolve the dry enzyme in a little water before charging. The reaction mixture is then heated to the gelatinization temperature and held there (or slightly hotter) until the required viscosity is reached. Potato, tapioca, sago, arrowroot, and cereal starches other than those obtained from maize or rice are gelatinized at 70 - 75 °C. Maize and rice starches require a temperature of 80 °C. When the conversion is complete, the mixture is heated quickly to 100 °C and held 10 minutes to inactivate the enzyme. Additional heating at 100 °C is sometimes done to effect further reduction in viscosity by the action of heat alone.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty