Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetyl Coenzyme A Trilithium Salt Trihydrate

Supplier: MP Biomedicals
0210049025 0210049005 0210049010 0210049001 0210049080
IC10049080EA 1010.67 USD InStock
IC10049080 IC10049005 IC10049025 IC10049001 IC10049010
Acetyl Coenzyme A Trilithium Salt Trihydrate
Enzymes
Acetyl-CoA is produced via beta-oxidation of fatty acids, via the metabolism of carbohydrates - glucose 6-phosphate to pyruvate to acetyl-CoA and via the catabolism of amino acids. Acetyl-CoA has a number of metabolic opportunities. It is metabolized in the tricarboxylic acid cycle to produce carbon dioxide, water and energy.

  • Extinction Coefficient (E1%): EM:(260 nm): 16400 (water), (259): 15400 (0.1 M PO₄ buffer, pH 7), (232 nm): 8700 (water) (Lit.)
  • Soluble in water (100 mg/ml - clear, colorless solution)
  • GHS symbol and signal word: GHS07 and warning

It can also be metabolized to fatty acids, cholesterol and steroid hormones. Acetyl-CoA also participates in a number of acetylation reactions, including the formation of acetylcholine, melatonin, N-acetylglucosamine, N-acetylgalactosamine and N-acetylneuraminic acid. Acetyl-CoA is involved in the acetylation of proteins and peptides. Histone acetylation is an epigenetic mechanism of gene regulation. In general, chromatin fractions enriched in actively transcribed genes are also enriched in highly acetylated core histones, whereas silent genes are associated with nucleosomes with a low level of acetylation. Nucleosomes are the fundamental units of chromosomes.

Acetyl coenzyme A is an essential cofactor and carrier of acyl groups in enzymatic acetyl transfer reactions. It is a key precursor in lipid biosynthesis, and the source of all fatty acid carbons. It is a positive regulator of pyruvate carboxylase. It is a percursor of the neurotransmitter acetylcholine.The acetic acid moiety which is bound by a high-energy bond (free energy 34.3 kJ/mol) to the -SH group of coenzyme A is a precursor to fatty acids, steroids and other naturally occurring compounds, such as terpenes and acetogenins present in plants.

The biosynthetic pathways for acetyl-CoA have been published. In the transfer reaction by acetyl-CoA of the C2 acetyl fragment, either the carboxyl group or the methyl group may react (electrophilic vs. nucleophilic reaction, respectively). Acetyl-CoA is an essential cofactor and carrier of acyl groups in enzymatic acetyl transfer reactions. It is formed either by the oxidative decarboxylation of pyruvate in mitochondria, by the oxidation of long-chain fatty acids, or by the oxidative degradation of certain amino acids. Acetyl-CoA is the starting compound for the citric acid cycle (Kreb′s cycle). It is also a key precursor in lipid biosynthesis, and the source of all fatty acid carbons. Acetyl-CoA positively regulates the activity pyruvate carboxylase. It is a precursor of the neurotransmitter acetylcholine. Histone acetylases (HAT) use Acetyl-CoA as the donor for the acetyl group use in the post-translational acetylation reactions of histone and non-histone proteins.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty