Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bovine Hyaluronidase (from Testes)

Supplier: MP Biomedicals
0210074091 0210074090 0210074080
IC10074091EA 408.38 USD InStock
IC10074091 IC10074080 IC10074090
Bovine Hyaluronidase (from Testes)
Enzymes
Hyaluronidase is a glycoprotein containing 5% mannose and 2.17% glucosamine, it catalyzes the random hydrolysis of 1,4-linkages between 2-acetamido- 2-deoxy- b-D-glucose and D-glucose residues in hyaluronate

  • Presentation: Tan Powder
  • Gene ID: 281838
  • Gene Symbol and Alisases: HYAL1, HYAL 2
  • Uniport Number: Q8SQG8
  • Extinction Coefficient (E1%): ~8 (Lit.)
  • Optimum pH 4.5 - 6 (Lit.)
  • Inhibitors: Fe2+ and Fe3+ are inhibitory as are Mn2+ and Cu2+, Sodium chloride acts as a stabilizer
  • Dissolve 10 mg hyaluronic acid in 25 ml 0.1 M sodium phosphate buffer: pH 5.3 with 0.15 M sodium chloride

Testicular hyaluronidase hydrolyzes the endo-N-acetylhexosaminic bonds of hyaluronic acid and chondroitin sulfuric acids A and C (but not B), primarily to tetrasaccharide residues. Monosaccharides are not liberated. Hyaluronidase is often used in conjunction with collagenase to dissociate the extracellular matrix between cells of animal tissue, in order to release viable cells for use in tissue culture. It may also be used to clarify synovial fluids in order to make cell counts possible.

Hyaluronidase degrades hyaluronan and has been found to be inappropriately regulated during cancer progression. These enzymes randomly cleave β-N-acetylhexosamine-[1→4] glycosidic bonds in hyaluronic acid, chondroitin, and chondroitin sulfates.

Hyaluronidase from bovine testes is a tetramer consisting of 4 equal subunits with a molecular mass of 14 kDa each.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty