Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lactase (from Aspergillus oryzae)

Supplier: MP Biomedicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

0210078001 0210078080 0210078025 0210078005 0210078083
IC10078001EA 51.75 USD InStock
IC10078001 IC10078025 IC10078005 IC10078080 IC10078083
Lactase (from Aspergillus oryzae)
Enzymes
Soluble in water; Insoluble in ethanol, acetone and isopropyl alcohol.

One unit will hydrolyze 1.0 µmole of o-nitrophenyl-beta-D-galactoside to o-nitrophenol per minute at pH 4.5 and 30 °C.

Stability: Stable in pH range of 4.0 to 8.0. Stable to heat. Optimum pH is 4.5 and 4.8 when O-Nitrophenyl-beta-D- galactopyranoside and Lactose are used as substrate, respectively(Lit.)

Lactase is an enzyme preparation produced by Aspergillus oryzae fermentation. Lactase hydrolyzes Lactose (4-o-β-D-Galactopyranosyl-D-glucose) and produces β-D-Galactose and a-D-Glucose.

Lactase is used to screen for blue white colonies in the MCS of various plasmid vectors in Escherichia coli or other bacteria. Its primary commercial use is to break down lactose in milk to make it suitable for people with lactose intolerance.

β-galactosidase cleaves lactose into its monosaccharide components, glucose and galactose. It also catalyses the transglycosylation of glucose into allolactose, the inducer of β-galactosidase, in a feedback loop.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty