Would you like to log in again?
Papain
Supplier: MP Biomedicals
0210092180 0210092150 0210092190 0210092191 0210092125
IC10092125EA 67.69 USD InStock
IC10092125 IC10092190 IC10092180 IC10092191 IC10092150
Papain
Enzymes
Papain is used in dissecting solutions
One unit will hydrolyze 1.0 µmole of N-alpha-benzoyl-L-arginine ethyl ester (BAEE) per minute at 25 °C and pH 6.2.
Papain is a sulfhydryl protease from Carica papaya latex.
Papain is a cysteine protease that cleaves peptide bonds of basic amino acids, leucine, or glycine. It is also hydrolyzes esters and amides.
It is used to produce Fab fragments of antibodies. It is used for cell dissociation since it has been shown to be more effective and less damaging with certain tissues. Papain has been used to isolate morphologically intact cortical neurons from postnatal rats. Limited papain digestion is used for structural studies of enzymes and other proteins. In addition, papain is used in red cell serology to modify the red cell surface to enhance or destroy the reactivity of many red cell antigens. Papain is useful for platelet serology studies. Papain has also been used in the enzymatic synthesis of amino acids, peptides, and other molecules.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty