Would you like to log in again?
Yeast, Bakers Dried Active, Solid, MP Biomedicals
Supplier: MP Biomedicals
0210140005 0210140001
IC10140001EA 107.75 USD InStock
IC10140001 IC10140005
Yeast, Bakers Dried Active, Solid, MP Biomedicals
Media for Microbiology Dehydrated Media for Microbiology
Baker's yeast is of the species Saccharomyces cerevisiae.
Yeast, or Baker's yeast is commonly used as a leavening agent in baking bread and bakery products, where it converts the sugars present in the dough into carbon dioxide and ethanol. Dried Active has been reported to asymmetrically reduce carbon carbon double bonds of nitroalkenes transforming them into intermediates used for the preparation of various biologically active compounds such as alkaloids, steroids. It can also be used to produce ethanol via fermentation.
Caution: Research Use Only (RUO).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty