Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(+)-Glutamine ≥99%, white crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms: Q(S)-2,5-Diamino-5-oxopentanoic acidL-GlutamineLevoglutamide(S)-(+)-GlutamineGlnH-Gln-OH

0210180605 0210180690 0210180680 0210180625 0210180683
IC10180605EA 43.79 USD InStock
IC10180605 IC10180690 75875-584 IC10180680 IC10180683
L(+)-Glutamine ≥99%, white crystalline powder
L(+)-Glutamine
L-Glutamine, the uncharged and amidated analog of L-glutamic acid, is an important amino acid for the incorporation of NH4+ into biomolecules. It is biosynthesized from NH4+ and glutamate via the enzyme glutamate synthetase. In turn, degradation of glutamine to free the ammonia moiety is mediated by glutaminase. Glutamine also participates in acid-base regulation in vivo.

L-glutamine is an essential amino acid, which serves as a major energy source for cells in culture. L-glutamine is added for optimal cell performance in a culture medium ( Ghyslaine Vanier, (2007), Characterization of the invasion of porcine endothelial cells by Streptococcus suis serotype 2, Canadian Journal Of Camparative Medicine, 71(2), pages: 81-89). The utilization of glutamine under carbon starvation conditions by cultured Saccharomyces cerevisiae and its relation to the cytoplasmic compartmentation of the transcriptional activator Gln3 has been studied. The use of glutamine in cultured rat islets to probe a-ketoisocaproate oxidation has been investigated.

L-Glutamine is an essential amino acid that is a crucial component of culture media that serves as a major energy source for cells in culture. L-Glutamine is very stable as a dry powder and as a frozen solution. In liquid media or stock solutions, however, L-glutamine degrades relatively rapidly. Optimal cell performance usually requires supplementation of the media with L-glutamine prior to use.

Soluble in water 41300 mg/l (at 25 °C).

Caution: Research Use Only (RUO).


Formula: C₅H₁₀N₂O₃
MW: 146.15 g/mol
Boiling Pt: 355 °C (1013 hPa)
Melting Pt: 185 °C (lit.)
Density: 1.5326 g/cm³ (130 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008044
CAS Number: 56-85-9
EINECS: 200-292-1
Merck Index: 13,04484
https://us-legacy.vwr.com/stibo/web/std.lang.all/98/28/2299828.jpg
Order Now
Specification Test Results
Identity Test Passes
Purity ≥99%
Residue on Ignition 0.1%
FTIR Conforms to Standard
Moisture 0.5%
Optical Rotation +30 ± 5° (c = 3.5, 1N HCl)
pH (2% aq soln) 4.5-6.5

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty