Would you like to log in again?
Pig Lipase
Supplier: MP Biomedicals
0210218920 0210218990 0210218980
IC0210218920EA 49.53 USD InStock
IC0210218920 IC10218990 IC10218980
Pig Lipase
Enzymes
Lipases are a subclass of the esterases. It catalyzes the hydrolysis of emulsified esters of glycerol and long chain fatty acids.
  • Presentation: Cream Colored Powder
  • Extinction Coefficient (E1%): E1%280 = 13.3(Lit.)
  • Isoelectric point (pI): Lipase A = 4.9 and Lipase B = 5.0 (Lit.)
  • Solubility: Partly soluble in water and insoluble in alcohol.
Lipases are used industrially for the resolution of chiral compounds and the transesterification production of biodiesel.
Lipases catalyze the hydrolysis of esters in aqueous solutions and the synthesis of esters in non-aqueous solutions.They also produce hydroxyl and carboxylic groups through the hydrolysis of ester linkages in poly(ethylene terephthalate).
Lipases catalyze the hydrolysis of triacylglycerols into glycerol and free fatty acids.
One USP unit of lipase activity is contained in the amount of pancreatin that liberates 1.0 µEq of acid per minute at a pH of 9.0 at 37 °C under the conditions of the assay for lipase activity.
Lipases are a subclass of the esterases. It catalyzes the hydrolysis of emulsified esters of glycerol and long chain fatty acids.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty