Would you like to log in again?
Pig Pepsin
Supplier: MP Biomedicals
0210259890 0210259820 0210259850 0210259880
IC10259850EA 128.27 USD InStock
IC10259850 75875-594 IC10259890 IC10259880
Pig Pepsin
Enzymes
Pepsin, an acid protease, contains a proteolytic enzyme
  • Presentation: Off-White Lyophilized Powder
  • Extinction Coefficient (E1%): E1%280 = 14.7
  • Isoelectric point (pI): pH 1.0
  • pH: 2.5 to 4.5 (2% aq solution)
  • Freely soluble in water; dissolves readily in 0.01 M HCl (0.5 mg/mL - clear, colorless solution); practically insoluble in alcohol, chloroform or ether.
Pepsin is a peptidase used to digest proteins and is commonly used in the preparation of Fab fragments from antibodies. Pepsin, from porcine gastric mucosa, has been used to hydrolyze dry cervical samples in mice.
Pepsin hydrolyzes peptide bonds, not amide or ester linkages. Pepsin cleaves peptides with an aromatic acid on either side of the peptide bond. Sulfur-containing amino acids increase susceptibility to hydrolysis when they are close to the peptide bond. Pepsin preferentially cleaves at the carboxyl side of phenylalanine and leucine and at the carboxyl side of glutamic acid residues. Cleaves Phe-Val, Gln-His, Glu-Ala, Ala-Leu, Leu-Tyr, Tyr-Leu, Gly-Phe, Phe-Phe and Phe-Tyr bonds in the β chain of insulin. Pepsin is the major proteolytic enzyme produced in the stomach. It digests proteins through the cleavage of interior peptide linkages.
Pepsin contains the "cathepsin" component which has milk curdling activity. It has a broad range of substrate activity and demonstrates an esterase acitivity. It generally attacks peptide bonds.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty