Would you like to log in again?
Poly-L-lysine hydrobromide
Supplier: MP Biomedicals
0210269180 0210269125 0210269191 0210269190
IC10269190EA 1040.38 USD InStock
IC10269190 IC10269180 IC10269191 IC0210269125
Poly-L-lysine hydrobromide
Proteins and Peptides
Poly-L-lysine is a positively charged amino acid polymer. Poly-lysine binds to DNA, red cell membrane and any negatively charged protein. It is typically used as a coating substrate for culture dishes, slides, etc. It enhances electrostatic interaction between negatively charged ions of the cell membrane and the culture surface. When adsorbed to the culture surface, poly-lysine increases the number of positively charged sites available for cell binding.
Other applications of Poly-L-lysine include Conjugation to methotrexate for increased drug transport, Microencapsulation of islets, Use in simple reproducible procedure for chromosomal preparations from a variety of tissues, Immobilation of living cells in biocompatible semipermeable microcapsules, In the preparation of polycationic beads, Conjugation to albumin and horseradish peroxidase to enhance cellular uptake.
Poly-L-lysine is a nonspecific attachment factor for cells useful in promoting cell adhesion to solid substrates by enhancing electrostatic interaction between negatively charged ions of the cell membrane and the culture surface. When it is absorbed to the cell culture surface, poly-L-lysine functions to increase the number of positively charged sites available for cell binding. With cells that can digest poly-L-lysine, poly-D-lysine should be used as the attachment factor.
There is approximately one HBr or HCl per lysine unit. The HBr/HCl allows it to be a crystalline solid and soluble in water. To remove the HBr/HCl, solubilize the product in a neutral buffer and dialyze away the salts.
Soluble in water (50 mg/ml-clear, colorless solution)
Caution: Research Use Only (RUO).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty