Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Soy Hydrolysate, MP Biomedicals

Supplier: MP Biomedicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

0210294105 0210294101
IC10294101EA 94.14 USD InStock
IC10294101 IC10294105
Soy Hydrolysate, MP Biomedicals
Media for Microbiology Additives and Supplements for Microbiology
Soy hydrolysate is an enzymatic (papain) digest of soybean meal. It is a plant source peptone. Soy hydrolysate is useful in media where high clarity is not required.

  • Yellowish tan powder
  • pH (2% aq soln): 6.7 to 7.5
  • Sulphated ash content ≤11.5%
  • Store at room temperature (15 to 30 °C)

Soluble in 100 mg/ml water. Loss of weight on drying is ≤5%. It contains total nitrogen- ≥8.0%; nitrogen, AN/TN - 17.0 to 28.0. Standard plate count to be ≤10000 and Enterobacteriaceae ≤10 CFU/G.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty