Would you like to log in again?
Trypsin
Supplier: MP Biomedicals
0210313990 0210313991 0210313925 0210313980 0210313983
IC10313925EA 157.68 USD InStock
IC10313925 IC10313990 IC10313991 IC10313980 IC10313983
Trypsin
Enzymes
Soluble in water; practically insoluble in alcohol and glycerol.
One USP trypsin unit is the activity causing a change in absorbance of 0.003 per minute under the conditions specified.
The protease activity of trypsin is highly specific toward positively charged side chains with lysine and arginine. Forms complexes with a2-macroglobulin.Can be used in the isolation of intact, detergent-free phycobilisomesand in the hydrolysis/condensation of carboxylic ester bonds.
Inhibitors: Trypsin is inhibited by organophosphorus compounds such as diisopropyl fluorophosphate and natural trypsin inhibitors from pancreas, soybean, lima bean and egg white. Silver ions are also potent inhibitors. Specific inhibitors are AEBSF, antipain, aprotinin, DFP, leupeptin, PMSF, TLCK, and Trypsin Inhibitor. Solutions: Solutions of trypsin should be thawed and swirled gently to mix. Solutions are relatively stable for periods of up to three months under refrigeration. If longer storage is desired, the concentrate should be aliquoted and refrozen. Thawed concentrate may contain a small amount of precipitate; this is normal and in no way will affect the efficacy of the product.
Trypsin is used to re-suspend cells adherent to the cell culture dish wall during the process of harvesting cells. Trypsin can also be used to dissociate dissected cells. Trypsin can be used to break down casein in breast milk. Trypsin is commonly used in biological research during proteomics experiments to digest proteins into peptides for mass spectrometry analysis. Trypsin can also be used to dissolve blood clots in its microbial form and treat inflammation in its pancreatic form.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty