Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Yeast Extract, Yellow-Tan Powder, MP Biomedicals

Supplier: MP Biomedicals
0210330380 0210330390 0210330391
IC10330390EA 140.81 USD InStock
IC10330390 IC10330380 IC10330391
Yeast Extract, Yellow-Tan Powder, MP Biomedicals
Media for Microbiology Dehydrated Media for Microbiology
Yeast Extract Powder is a vacuum-dried extract concentrate of Baker's yeast containing the B-complex factors of approximately three times the weight of ordinary dry yeast. This product is a vacuum dried autolyzed yeast extract produced from 100% Baker's yeast. It is a concentrated source of soluble protein components, carbohydrates, and B Complex vitamins natural to yeast. Water soluble portion of autolyzed yeast with intact B-complex vitamins. Yeast extract is a mixture of amino acids, peptides, water soluble vitamins and carbohydrates and can be used as additive for culture media.
Yeast Extract Powder is a highly nutritive medium recommended for plate count of microorganisms in water. It is used as a bacteriological media component for a variety of microorganisms and molecular genetics applications. Also suitable for alternative to beef extract.

  • Suitable for alternative to beef extract
  • Presentation: Yellow-Tan Powder
  • pH: 5-5.5 (aq.soln) (Lit.)
  • Storage temperature: Room Temperature, desiccate
  • Solubility (5g/100ml): Clear
  • Moisture: <5%
  • Mold Count: < 50/gram
  • Standard Plate Count: < 50,000/gram
  • Salmonella: Negative

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty