Would you like to log in again?
Cholesterol Esterase (from Pseudomonas sp.), MP Biomedicals
Supplier: MP Biomedicals
0210543950 0210543982 0210543991
IC10543991EA 885.42 USD InStock
IC10543991 IC10543982 IC10543950
Cholesterol Esterase (from Pseudomonas sp.), MP Biomedicals
Enzymes
Excess cholesterol is stored intracellularly as cholesterol esters.
  • Presentation: Yellowish tan lyophilized powder
  • Isoelectric point (pl): 5.9 ±0.1(Lit.)
  • Optimum pH: 7.0 to 9.0 ; pH stability: pH 5.0 to 9.0 (25 °C, 24 h)
  • Soluble in distilled water or dilute buffer
Cholesterol Esterase is useful for enzymatic determination of total cholesterol when coupled with cholesterol oxidase in clinical analysis.
Cholesterol esterase (CE) is a reversible enzyme that can hydrolyze or synthesize fatty acid esters of cholesterol and other sterols. Hydrolysis of water insoluble long chain fatty acid esters requires bile salt activation. Hydrolysis of water soluble esters of short chain fatty acids and lysophospholipids does not require activation by bile salts. It also hydrolyzes tri-, di-, and mono-acylglycerols, phospholipids, lysophospholipids, and ceramide. The enzyme may have multiple functions in lipid and lipoprotein metabolism, and atherosclerosis.
Cholesterol esterase (CE) is a reversible enzyme that can hydrolyze or synthesize fatty acid esters of cholesterol and other sterols.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty