Would you like to log in again?
SOC Powder, MP Biomedicals
Supplier: MP Biomedicals
113031012
IC113031012EA 87.44 USD InStock
IC113031012
SOC Powder, MP Biomedicals
Media for Microbiology Dehydrated Media for Microbiology
SOC Medium is a rich media used primarily in the recovery step of Escherichia coli competent cell transformations.
  • Special medium for incubating cells immediately after transformation
  • Allows expression of transferred resistance genes before exposing cells to selective conditions
  • Maximizes the transformation efficiency of competent cells
Reagents are tested and chosen for their ability to promote optimum growth of E. coli when combined in SOC formulations. Chosen reagents are combined in the proper proportions according to the SOC formulation and thoroughly blended for a uniform distribution. A sample of the blended formulation was used to prepare liquid medium by adding 31 g of powder per liter of purified water. The combinations were mixed by hand for about 1 minute to dissolve and autoclaved at 121 °C for 15 minutes. Autoclaving at higher temperatures or for longer periods will result in darkening of the sterile medium, (a darker color does not noticeably affect growth of cells, only the appearance of the sterile medium).
Ordering information: Small quantities of biological buffers have been added for proper pH adjustment.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty