Would you like to log in again?
CSM-Arg Powder, MP Biomedicals
Supplier: MP Biomedicals
114510122 114510112
IC114510122EA 1100.14 USD InStock
IC114510122 IC114510112
CSM-Arg Powder, MP Biomedicals
Media for Microbiology Dehydrated Media for Microbiology
Single dropout of Complete Supplement Mixture (CSM) of amino acids for S. cerevisiae growth media.
  • Transcription Regulation
  • Time tested formulation is available in complete and dropout formulations
  • For use with DOB or YNB with the appropriate nitrogen and carbon sources to custom formulate your own selective Synthetic Defined yeast media
Single Dropout Complete Supplement Mixture used to select for yeast containing plasmids with specific nutritional markers and to select for gene function in transformation experiments.
Reagents are tested and chosen for their ability to promote optimum growth of yeast strains when combined in the DOB or DOBA with CSM-Arg formula. DOB (Drop Out Base) is 1.7 g Yeast Nitrogen Base, 5 g Ammonium Sulfate, 20 g Dextrose per liter. DOBA is DOB with 17% agar. Chosen reagents are combined in the proper proportions according to the formulation listed in the certificate of analysis and milled to a powder which thoroughly blends the reagents for uniform distribution. A sample of the milled and blended formulation was used to prepare liquid plate medium by adding 43.7 g DOBA powder and 0.74g of CSM-Arg to a liter of purified water. The combinations were mixed by hand for about 1 minute to dissolve the dextrose and autoclaved at 121 °C for 25 minutes. After cooling to 50 °C plates were poured.
Certifications: Molecular Biology Certified™
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty