Would you like to log in again?
BipPulverizer System II, MP Biomedicals
Supplier: MP Biomedicals
116850200 116850200
IC116850200EA 118.01 USD InStock
IC116850200 CA76046-292
BipPulverizer System II, MP Biomedicals
Buffers Extraction Buffers
BioPulverizer™ System II contains 10x2 mL tubes of each of our specialized Lysing Matrices.
  • Contains Matrix types F, G, H, I, and J
  • This sampler pack gets you going
  • Optimizes homogenization
  • Ideal for very difficult, unusual, or multiple samples
Contains 10 each of the impact-resistant 2 mL tube of: Lysing Matrix F, Lysing Matrix G, Lysing Matrix H, Lysing Matrix I, and Lysing Matrix J.
Matrix F contains 1.6 mm aluminum oxide granules and 1.6mm silicon carbide granules. Matrix G contains 1.6mm silicon carbide granules and 2 mm glass beads. Matrix H contains 2 mm glass and 2 mm yellow zirconium oxide beads. Matrix I contains 2 mm yellow zirconia beads and a 4 mm black ceramic bead. Matrix J contains 2 mm yellow zirconia beads and 1.6mm aluminum oxide granules.
Certifications: DNase and RNase Free
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty