Would you like to log in again?
beta-Galactosidase
Supplier: MP Biomedicals
0215003901 0215003903
IC15003903EA 453.27 USD InStock
IC15003903 IC15003901
beta-Galactosidase
Enzymes
One unit catalyzes the hydrolysis of 1 µmole of o-nitrophenyl-β-D-galactopyranoside per minute at 25 °C, pH 7.5.
Substrate Specificity: The enzyme specifically hydrolyzes β-D-galactosyl linkage
Inhibitors: p-Chloromercuribenzoate, lodoacetamide, heavy metal ions (Zn++, Fe++, Zn++, Cd++, Cu++, Pb++, Ag+, Hg++), Ionic Detergents (SDS, DAC, etc.). Contaminants: The preparation is practically free from other glycosidases (a-galactosidase, a-,b-glucosidase, a-,b-mannosidase, etc.) and proteinase. Principle: o-Nitrophenyl-b-D-galactopyranoside (ONPG) b-galactosidase > o-Nitrophenol (ONP) + D-Galactose. The appearance of o-nitrophenol is measured at 410 nm by spectrophotemetry. Thermal Stability: Below 50 °C (pH 7.3, 15 min) (Lit.), Optimum Temperature: 50 - 55 °C (Lit.)
β-galactosidase cleaves lactose into its monosaccharide components, glucose and galactose. It also catalyses the transglycosylation of glucose into allolactose, the inducer of β-galactosidase, in a feedback loop.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty