Would you like to log in again?
Amylase
Supplier: MP Biomedicals
02150375.2 0215037501 0215037505
IC15037505EA 837.51 USD InStock
IC15037505 IC150375.2 IC15037501
Amylase
Enzymes
The number of grams of soluble starch dextrinized in the presence of excess B-amylase per hour at 40 °C, pH 5.0.
  • Optimal temperature 50 to 55 °C
  • Activated by Ca2+ (Lit.)
The number of grams of soluble starch dextrinized in the presence of excess B-amylase per hour at 40 °C, pH 5.0.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty