Would you like to log in again?
Cellulase (from Aspergillus niger)
Supplier: MP Biomedicals
0215058305 0215058325 0215058380
IC15058305EA 51.75 USD InStock
IC15058305 IC15058325 IC15058380
Cellulase (from Aspergillus niger)
Enzymes
Cellulase refers to a group of enzymes which, acting together, hydrolyze cellulose
  • Presentation: Tan Powder
  • Optimum pH: Varies with the substrate in the range 4.2 - 5.2.(Lit.)
  • Readily soluble in water
Cellulase is used for commercial food processing in coffee. It performs hydrolysis of cellulose during drying of beans. Furthermore, cellulases are widely used in textile industry and in laundry detergents. They have also been used in the pulp and paper industry for various purposes, and they are even used for pharmaceutical applications. Cellulase is used in the fermentation of biomass into biofuels, although this process is relatively experimental at present. Cellulase is used as a treatment for phytobezoars, a form of cellulose bezoar found in the human stomach.
Cellulase from Aspergillus niger catalyzes the hydrolysis of endo-1,4-β-D-glycosidic linkages in cellulose, lichenin, barley glucan, and the cellooligosaccharides cellotriose to cellohexaose. It does not cleave cellobiose or p-nitrophenyl-β-D-glucoside. This enzyme will also cleave intact glycosaminoglycan from a core peptide by hydrolyzing the xylosyl serine linkage.
The b-glucosidases, including cellobiose, are primarily active on the smaller molecular weight cellulose hydrolysates. During cellulose breakdown they are active on the dimers and oligomers of cellulose.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty