Would you like to log in again?
Collagenase Type 3
Supplier: MP Biomedicals
0215070401 0215070405
IC15070405EA 1885.1 USD InStock
IC15070405 IC15070401
Collagenase Type 3
Enzymes
For typical assay, dissolve enzyme at a concentration of 1 mg/mL in 0.05 M TES with 0.36 mM calcium chloride, pH 7.5
One unit liberates one µmole of L-leucine equivalents from collagen in 5 hours at 37 °C, pH 7.5.
Dilutions run are 1/10 and 1/20 in the above buffer.
Activators: Ca2+ is required. ; Inhibitors: Metal chelating agents such as cysteine, EDTA or o-phenanthroline but not DFP. It is also inhibited by a2-macroglobulin.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty