Would you like to log in again?
Collagenase
Supplier: MP Biomedicals
0215070501 0215070505
IC15070505EA 2555.39 USD InStock
IC15070505 IC15070501
Collagenase
Enzymes
Collagenase can be solubilized in calcium-free solutions such as Hank's Balanced Salts or Earles Balanced Salt Solution.
Unit definition: One unit liberates one µmole of L-leucine equivalents from collagen in 5 hours at 37 °C, pH 7.5.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty