Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Glutamic Dehydrogenase

Supplier: MP Biomedicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

0215118980 0215118990 0215118950
IC15118950EA 100.14 USD InStock
IC15118950 IC15118990 IC15118980
L-Glutamic Dehydrogenase
Enzymes
For Continuous Spectrophotometric Rate Determination method: 0.3 to 0.6 unit/ml of L-Glutamic dehydrogenase in cold Triethanolamine buffer.

One unit will cause the transformation of 1.0 µmole of alpha-ketoglutarate per minute at pH 7.3 and 25 °C.

LDH ≤0.01% ; MDH ≤0.01% ; Inhibitor: Dimethyl dicarbonate.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty