Would you like to log in again?
Hyaluronidase
Supplier: MP Biomedicals
0215127080
IC15127080EA 195.31 USD InStock
IC15127080
Hyaluronidase
Enzymes
Unit Definition: Enzyme activity is determined by a change in optical density in a solution of hyaluronic acid. Activity is compared to a hyaluronidase International Standard and is expressed as an International Unit (U).
  • Optimum pH: 6.0
  • Recommended reaction temperature: 60 °C
  • Inhibitors: Mn2+, Hg2+, Potassium ferricyanide.
Hyaluronidase purified from Streptomyes hyalurolyticus is distinct from the known hyaluronidase in its substrate specificity and mechanism of action. This enzyme is quite specific for hyaluronic acid. Various types of chondroitin sulfate (A,B,C,D,,E), chondroitin, heparin, keratan sulfate and chitin are not attacked by this enzyme.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty