Would you like to log in again?
Hyaluronidase
Supplier: MP Biomedicals
0215127480 0215127425
IC15127480EA 456.11 USD InStock
IC15127480 IC15127425
Hyaluronidase
Enzymes
Dissolve 10 mg hyaluronic acid in 25 mL 0.1 M sodium phosphate buffer: pH 5.3 with 0.15 M sodium chloride.
Amount of enzyme which liberates one micromole of N-acetylglucosamine per minute at 37°C and pH 4.0
Inhibitors: Fe2+ and Fe3+ are inhibitory as are Mn2+ and Cu2+, Stabilizers: Sodium chloride acts as a stabilizer.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty