Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Protease (from S. aureas V8 strain)

Supplier: MP Biomedicals
0215197201 0215197205 0215197210
IC15197210EA 1840.64 USD InStock
IC15197210 IC15197201 IC15197205
Protease (from S. aureas V8 strain)
Enzymes
Protease from Staphylococcus aureus strain V8 is composed of a single polypeptide chain

  • Presentation: Off-white Powder
  • Extinction Coefficient (E1%): E1%280 = 4.26
  • pH: 4.0 and 7.8 with hemoglobin substrate (optimum pH)
  • Inhibitors: Diisopropyl fluorophosphate (DFP) and monovalent anions such as F-, Cl-, Br-, CH3COO- and NO3-.
  • Soluble in aqueous buffers (such as 10 mM Tris, pH 7.8 at 1 mg/mL).

Protease V8 is used for selective cleavage of proteins for amino acid sequence determination or peptide mapping.

The enzyme was isolated, by the method of Drapeau et al., from the culture filtrate of S. aureus V8 strain. The preparation is homogenous on ultracentrifugation and disc electrophoresis.

The Staphylococcus strain V8 protease specifically cleaves peptide bonds on the carboxyl (COOH) terminal side of aspartic and glutamic acid residues.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty