Would you like to log in again?
Alpha-Chymotrypsin
Supplier: MP Biomedicals
02152272.2 0215227205 0215227201
IC15227201EA 153.74 USD InStock
IC15227201 IC15227205 IC152272.2
Alpha-Chymotrypsin
Enzymes
One unit will hydrolyze 1.0 µmole of N-benzoyl-L-tyrosine ethyl ester (BTEE) per minute at pH 7.8 and 25 °C in the presence of ionized calcium.
Inhibitors: The enzyme is inhibited by heavy metals, the natural trypsin inhibitors to various degrees, an inhibitor from potato, and organophosphorus compounds. Gel filtration of chymotrypsin removes autolysis products and other contaminants. The specificity of α-chloroketone as α-chymotrypsin inhibitor has been studied. It has been reported that phenothiazine-N-carbonyl chloride are specific for chymotrypsin inhibition. Also inhibited by AEBSF, α-1-antitrypsin, Aprotinin, DFP, PMSF, TPCK and α-2-Macroglobulin.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty