Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eosin Y (yellowish), red powder C.I. 45380

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning
Synonyms: Tetrabromofluorescein disodium saltEosin Y disodium saltEosin 2252′,4′,5′,7′-Tetrabromofluorescein disodium saltSolvent Red 43Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoateDisodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate)Field's Stain B2,4,5,7-Tetrabromofluorescein disodium salt

0215247183 0215247180 0215247125
IC15247183EA 261.76 USD InStock
IC15247183 IC15247180 IC15247125
Eosin Y (yellowish), red powder C.I. 45380
Eosin Y (yellowish)
Counterstain for hematoxylin.

Eosin Y disodium salt is used in combination with Azure II to show morphological details of marrow cells; buffered Azure A/Eosin Y method for demostrating cell granules, microorganisms and nuclei; for showing insect musculature; for demonstrating cells infected with mosquito-iridescent virus; use in combination with Alum Hematoxylin/Azure II to show mast cells, early necrobiotic changes in cells and protozoans, rickettsias and bacteria in tissues; to demonstrate volutin granules in any microorganism; Bond's modification of Mann's Eosin-Methyl Blue for Negri bodies in smears; Harris stain for Negri bodies in smears; Mann's method for staining Negri bodies in sections; Gridley's stain for Endamoeba in tissue sections; a modified methenamine-silver nitrate method for the demonstration of Endamoeba in sections. It is used as a counterstain for hematoxylin.

Soluble in water (40 mg/mL); ethanol (10 mg/mL); Ethylene Glycol Monomethyl Ether (Methyl-cellosolve; EGME) (50 mg/mL).

Caution: Research Use Only (RUO).


Formula: C₂₀H₆Br₄Na₂O₅
MW: 691.86 g/mol
MDL Number: MFCD00005040
CAS Number: 17372-87-1
Order Now
Specification Test Results
UV Visible Absorbance λ max 515 - 518 nm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty