Would you like to log in again?
Pyruvate Oxidase
Supplier: MP Biomedicals
0215348625
IC15348625EA 86.22 USD InStock
IC15348625
Pyruvate Oxidase
Enzymes
One unit causes the formation of one µmole of hydrogen peroxide (half a µmole of quinoneimine dye) per minute at pH 5.7 at 37 °C.
Michaelis constant : 3.4 X 10^-4M (Pyruvate) Inhibitors : Fe++,Zn++,Cu++,Ag+,Hg++ Optimum temperature : 65°C, pH Stability : pH 5.7 - 6.5 (25°C,, 20hr) Thermal stability : below 45°C, (pH 6.0, 15min)
Pyruvate Oxidase reacts with certain aldehydes and phosphate can be replaced by arsenate. Oxygen as well as several artificial compounds can function as electron acceptors. Pyruvate Oxidase is activated by phospholipids as well as monomeric and micellar amphiphiles.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty