Would you like to log in again?
Choline Oxidase, MP Biomedicals
Supplier: MP Biomedicals
0215349250
IC15349250EA 89.74 USD InStock
IC15349250
Choline Oxidase, MP Biomedicals
Enzymes
Dilute in 10 mM Tris-HCl buffer, pH 8.0 containing 2 mM EDTA and 1.0% KCl.
One unit causes the formation of one µmole of hydrogen peroxide per minute at pH 8.0 at 37 °C.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty