Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide ≥97%, pellets

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

0215349501 02153495.5
IC15349501EA 140.2 USD InStock
IC15349501 IC153495.5
Sodium hydroxide ≥97%, pellets
Sodium hydroxide
1g dissolves in 0.9 mL water; 0.3 mL boiling water, 7.2 mL absolute alcohol, 4.2 mL methanol; insoluble in glycerol. The dissolution of sodium hydroxide in water is a highly exothermic reaction. Sodium hydroxide stock solutions should be stored in plastic containers. Glass containers should be avoided in the preparation and storage of sodium hydroxide solutions because it will etch the glass (typically anything over 2 N will etch glass). Keep solutions protected from air as much as possible since contact with air will form sodium carbonate.

Poison/Deleterious Substance Code: 2-54


Formula: NaOH
MW: 40 g/mol
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1823
ADR: 8,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/99/42/36399942.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty