Would you like to log in again?
Bovine Thrombin
Supplier: MP Biomedicals
0215416301 0215416380 0215416350 0215416310 0215416305
IC15416310EA 313.36 USD InStock
IC15416310 IC15416380 IC15416350 IC15416305 IC15416301
Bovine Thrombin
Enzymes
Thrombin is prepared from fresh bovine plasma by an ion exchange method
  • High Purity Grade
  • Concentration: ~10% (Protein)
  • Presentation: White Powder
  • Plasminogen and plasmin not detected.
α-Thrombin is a trypsin-like serine protease involved in a multitude of processes in the human body. Thrombin generation is the result of limited proteolysis of the vitamin K-dependent zymogen prothrombin. Thrombin is the last enzyme in the clotting cascade functioning to cleave fibrinogen to fibrin which forms the fibrin gel of a hemostatic plug or a pathologic thrombus. In addition, thrombin potentiates the procoagulant process by activating Factors V, VIII, XI and XIII.
Serine protease that selectively cleaves Arg-Gly bonds in fibrinogen to form fibrin and fibrinopeptides A and B.
Fibrinogen is removed from the prothrombin complex by precipitation. The concentrated prothrombin complex is then activated by beef thromboplastin and calcium. The thrombin solution is clarified by centrifugation and filtration and then lyophilized.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty