Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Patton-Reeders reagent (Calconcarboxylic acid), Powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms: 2,2'-Dihydroxy-4'-sulpho-1,1'-azonaphthalene-3-carboxylic acid3-Hydroxy-4-(2-hydroxy-4-sulpho-1-naphthylazo)naphthalene-2-carboxylic acid2-Hydroxy-1-(2-hydroxy-4-sulpho-1-naphthylazo)-3-naphthoic acid2-Hydroxy-1-(2-hydroxy-4-sulfo-1-naphthylazo)-3-naphthoic acidCal-Red®3-Hydroxy-4-(2-hydroxy-4-sulfo-1-naphthylazo)naphthalene-2-carboxylic acid2,2'-Dihydroxy-4'-sulfo-1,1'-azonaphthalene-3-carboxylic acid

0215745105
IC15745105EA 47.64 USD InStock
IC15745105
Patton-Reeders reagent (Calconcarboxylic acid), Powder
Patton-reeders reagent (Calconcarboxylic acid)
2-Hydroxy-1-(2-hydroxy-4-sulfo-1-naphthylazo)-3-naphthoic acid is a useful indicator for calcium titrations with EDTA in the presence of magnesium.

Soluble in methanol (clear reddish violet solution), 50 mg plus 5 ml of 1 M NaOH; slightly soluble in water.


Formula: C₂₁H₁₄N₂O₇S
MW: 438.42 g/mol
MDL Number: MFCD00004078
CAS Number: 3737-95-9
Order Now
Specification Test Results
Identity Test Passes

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty