Would you like to log in again?
Alkaline Phosphatase
Supplier: MP Biomedicals
0215934705 0215934710
IC15934710EA 821.36 USD InStock
IC15934710 IC15934705
Alkaline Phosphatase
Enzymes
Dissolves readily at 5 mg/mL in a 0.025M glychine/NaOH pH 9.6 containing 1 mM magnesium chloride, 0.1 mM zinc chloride and 10% glycerol, to give a clear pale pink solution also soluble in distilled water or dilute buffer.
One unit liberates one µmole of p-nitrophenol per minute at 37 °C, pH 10.15 in Tris buffer. The enzyme activity is determined by measuring the increase in optical density at 405 nm resulting from the hydrolysis of p-nitrophenyl phosphate.
Activators: Zn, Mg2+, Ca2+, Tris buffer, diethanolamine; Inhibitors: Acidification to pH 4.5 reversibly inactives the enzyme; inorganic phosphate, chelating agents such as EDTA. Note: Alkaline phosphatase is fully active in the presence of sodium azide (0.05%) and Triton X-100 (0.1%).
Alkaline phosphatase is used for removing phosphate monoester to prevent self ligation. It can be also be used as a label for enzyme immunoassays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty