Would you like to log in again?
Aldolase
Supplier: MP Biomedicals
0215985925 0215985980
IC15985925EA 123.47 USD InStock
IC15985925 IC15985980
Aldolase
Enzymes
One unit is the change in absorbancy of 1.00 per minute at 25 °C, pH 7.5.
Inhibitors: Aldolase in inhibited by Cu2+, Ag+, Zn2+ and o-phenanthroline. It is inactivated by N-bromoacetylethanolamine phosphate5 and pyridoxal phosphate. Stability: Aldolase is irreversibly denatured at pH values less than 4.5.
Aldolase is a tetrameric protein. It catalyzes a key reaction in glycolysis and energy production:D-Fructose 1,6-bisphosphate Aldolase> dihydroxyacetone phosphate + D-glyceraldehyde-3-phosphate. Aldolase is present in all animal tissue and in most microorganisms. There are two classes of aldolases. Class I aldolase is found in animal and higher plant tissue. Class II aldolase is found in primitrive cells such as yeasts and bacteria. Class I aldolase is characterized by not requiring a bivalent metal cofactor and the formation of a ketimine Schiff base intermediate with the substrate dihydroxyacetone phosphate. Class II aldolase requires a metal cofactor and is inhibited by EDTA. Three types of aldolase exist in animal tissue. The major form, type A is found in muscle; type B is found in liver tissue and type C (plus some type A) is found in brain tissue. Aldolase forms five isozymes which may to various degrees be organ specific.
Aldolase is used in an hydrazine assay based on Boyer's modification in which 3-phosphoglyceraldehyde reacts with hydrazine to form a hydrazone which absorbs at 240 nm.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty