Would you like to log in again?
Bovine Superoxide Dismutase (from Erythrocytes)
Supplier: MP Biomedicals
0219011732 0219011703 0219011715 0219011775 0219011730
IC19011715EA 105.2 USD InStock
IC19011715 IC19011703 IC0219011732 IC19011775 IC19011730
Bovine Superoxide Dismutase (from Erythrocytes)
Enzymes
Superoxide dismutases are enzymes that catalyze the dismutation of superoxide into oxygen and hydrogen peroxide
  • Presentation: Pale Blue Powder
  • Extinction Coefficient (E1%): 2.64 (1%/280) (Lit.)
  • Isoelectric point (pl): 4.95 (Lit.)
  • Dissolves readily at 5mg/mL in 0.05M potassium phosphate/0.1mM EDTA pH 7.8, to give a clear slightly blue solution.
Superoxide Dismutase plays a critical role in the defense of cells against the toxic effects of oxygen radicals. Superoxide dismutase from bovine erythrocytes has been used in a study to assess a kinetic model of radiation-induced inactivation of superoxide dismutase in nitrous oxide-saturated solutions. Superoxide dismutase from bovine erythrocytes has also been used in a study to investigate the possible participation of superoxide anion in the intestinal tryptophan 2,3-dioxygenase reaction.
Superoxide dismutase catalyzes the dismutation of superoxide radicals to hydrogen peroxide and molecular oxygen. It competes with nitric oxide (NO) for superoxide anion (which reacts with NO to form peroxynitrite), thereby SOD promotes the activity of NO. SOD has also been shown to suppress apoptosis in cultured rat ovarian follicles, neural cell lines, and transgenic mice.
Thus, they are an important antioxidant defense in nearly all cells exposed to oxygen. Superoxide dismutases are a group of low molecular weight metalloproteins present in all aerobic cells of plants, animals and micro-organisms. They provide protection against damaging reactions with the superoxide radical anion (O2-) by catalyzing its disproportionation into oxygen and hydrogen peroxide.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty