Would you like to log in again?
Aldehyde Dehydrogenase (from Yeast)
Supplier: MP Biomedicals
0219067983 0219067925 0219067980
IC19067980EA 255.93 USD InStock
IC19067980 IC19067983 IC19067925
Aldehyde Dehydrogenase (from Yeast)
Enzymes
Aldehyde dehydrogenase belongs to the family of oxidoreductases, specifically those acting on the aldehyde or oxo group of donor with NAD+ or NADP+ as acceptor.
  • Presentation: Off-White Powder
  • Extinction Coefficient (E1%): 9.4 (279 nm)(Lit)
  • Optimum pH: 9.0
  • Gene ID: 855205
  • Uniport Number: P54114
Aldehyde dehydrogenase oxidizes a number of aliphatic and aromatic aldehydes. Acetyl-GSH is not hydrolyzed.
Aldehyde dehydrogenase is a tetramer and has several different isoforms. The enzyme tested in 0.01 M pyrophosphate buffer shows a sharp optimum around pH 9.3 with acetaldehyde as substrate. Potassium ions and cysteine are essential for the enzyme′s activity. Rubidium or NH4+ can be substituted for K+, and glutathione for cysteine. Lithium, Na+, and Cs+ inhibit the reaction. Aldehyde dehydrogenase is inhibited by propylurea, crotonaldehyde, n-propyl isocyanate, cyclohexyl isocyanate, 1-n-propyl-1-[(4-chlorophenyl)sulphonyl]-3-n-propylurea, and 1-methyl-1-[(4-chlorophenyl)sulphonyl]-3-n-propylurea. The enzyme may be utilized to quantitate aldehydes present in blood.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty