Would you like to log in again?
Alpha-Amylase
Supplier: MP Biomedicals
0219123910 0219123925
IC19123925EA 483.13 USD InStock
IC19123925 IC19123910
Alpha-Amylase
Enzymes
One unit catalyzes the liberation of 1 mmole reducing group (calculated as maltose per minute at pH 6.9 at 20 °C).
  • Origin: Product is of animal origin
One unit catalyzes the liberation of 1 mmole reducing group (calculated as maltose per minute at pH 6.9 at 20 °C).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty