Would you like to log in again?
Bovine IgG Isotype Control
Supplier: MP Biomedicals
0219147825
IC19147825EA 475.59 USD InStock
IC19147825
Bovine IgG Isotype Control
Antibodies
Soluble in normal saline solution (25 mg/mL, 0.9%NaCl).
Protein >90%
Bovine γ-globulin is a plasma protein fraction rich in antibodies.
Bovine γ-globulin has been used as an immunological challenge in studies of mechanisms of immunotolerance.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty