Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Proteinase K (from Tritirachium album)

Supplier: MP Biomedicals
0219350425 0219350490 0219350483 0219350491 0219350480
IC19350490EA 806.27 USD InStock
IC19350490 IC19350425 IC19350483 IC19350491 IC19350480
Proteinase K (from Tritirachium album)
Enzymes
A highly active stable endopeptidase with a broad spectrum of action was isolated by E.

  • Presentation: White to off white lyophilized powder
  • Isoelectric point (pI): 8.9
  • pH optimum (denatured hemoglobin as substrate): pH 7.5 to 12.00
  • Soluble in water (1 mg/ml)

Proteinase K is suitable for both protein and nucleic acid isolation. Exhibits proteolytic activity on proteins, peptides, glycoproteins, amides and esters. Also active with nitroanilides of amino acids with protected amino groups, excluding arginine. Useful in the isolation of DNA and RNA, in the analysis of membrane structures and protein structure. Addition of either 0.5 to 1% of sodium dodecyl sulfate or 1-4 mmol/L of urea increases the activity, because the substrates are more easily attacked when denatured. In experiment to recover the undigested DNA, proteins were digested with 0.1 mg/ml of proteinase K.

Proteinase K is a stable and highly reactive serine protease. Evidence from crystal and molecular structure studies indicates the enzyme belongs to the subtilisin family with an active-site catalytic triad (Asp39-His69-Ser224). It is stable in a broad range of environments: pH, buffer salts, detergents (SDS), and temperature. In the presence of 0.1 to 0.5% SDS, proteinase K retains activity and will digest a variety of proteins and nucleases in DNA preparations without compromising the integrity of the isolated DNA.

Proteinase K cleaves peptide bonds mostly after the carboxyl group of N-substituted hydrophobic aliphatic and aromatic amino acids, as shown by specificity trials with amino acid-4-nitroacilides. Thus, it shows similarities with alkaline Asperigillus proteases. However, unlike the latter, Protease K also cleaves peptide amides, comparable to the alkaline serine-proteases from Bacillus species. The specificity of ester cleavage is also high.

Merk's Darmstadt Biochemical Research Department in 1970 from a culture filtrate of the fungus, Tritirachium album Limber, suitable for both protein and nucleic acid isolation, it exhibits proteolytic activity on proteins, peptides, glycoproteins, amides and esters. Also active with nitroanilides of amino acids with protected amino groups, excluding arginine.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty