Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium chloride dihydrate ∼99%, white crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning
Synonyms: Calcium Chloride Dihydrate

0219508890
IC19508890EA 107.55 USD InStock
IC19508890
Calcium chloride dihydrate ∼99%, white crystalline powder
Calcium chloride dihydrate


Formula: CaCl₂·2H₂O
MW: 147.01 g/mol
MDL Number: MFCD00149613
CAS Number: 10035-04-8
Order Now
Specification Test Results
Identity Test Passes
Purity ~99%
pH (5% aq soln) 4 - 9.5

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty