Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium acetate, anhydrous ≥99.0%, crystalline powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms: Acetic acid sodium salt

0219549601 0219549602 02195496.5
IC19549601EA 196.51 USD InStock
IC19549601 IC195496.5 IC19549602
Sodium acetate, anhydrous ≥99.0%, crystalline powder ACS
Sodium acetate
Sodium acetate is used as a buffer in the buffering range of pH 3.6-5.6. It is also used in the purification and precipitation of nucleic acids, protein crystallization, staining of gels in protein gel electrophoresis, HPLC, mordant dyeing, DNA microarray studies of E. coli response, investigation of protein unfolding during reverse phase chromatography.

It is used in conjunction with sodium carbonate to enhance the activation of freeze-dried subtilisin Carlsberg in organic solvents. It is also used as substrate for acetokinase.

Soluble freely in water; moderately soluble in alcohol.


Formula: H₃CCOONa
MW: 82.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012459
CAS Number: 127-09-3
Order Now
Specification Test Results
Assay ≥99,0%
pH (5% aq soln) 7,0 - 9,2
Insoluble Matter ≤0,01%
Loss on Drying (120 °C) ≤1,0%
Chloride ≤0,002%
Phosphate ≤0,001%
Sulfate ≤0,003%
Calcium ≤0,005%
Magnesium ≤0,002%
Heavy Metals (as Pb) ≤0,001%
Iron ≤0,001%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty