Would you like to log in again?
Cell Biology Murashige and Skoog Basal Medium with Gamborg's Vitamins, MP Biomedicals
Supplier: MP Biomedicals
092623220
IC2623220CS 47.6 USD InStock
IC2623220
Cell Biology Murashige and Skoog Basal Medium with Gamborg's Vitamins, MP Biomedicals
Media for Microbiology Dehydrated Media for Microbiology
Murashige and Skoog medium is a widely used plant tissue culture growth medium.
  • Includes macro- and micronutrients
  • Plant Media
  • Soluble 1X in water
Formulation: Ammonium Nitrate 1650 mg/L 20.61 uM, Boric Acid 6.2mg/L 100 uM, Calcium Chloride, Anhydrous 332.2 mg/L 2.99 mM, Cobalt Chloride6H2O 0.025 mg/L 0.11 uM ,Cupric Sulfate5H2O 0.025 mg/L 0.1 uM, Na 2 -EDTA 37.26 mg/L 100 uM, Ferrous Sulfate7H2O 27.8 mg/L 100 uM, Magnesium Sulfate 180.7 mg/L 1.5 mM, Manganese SulfateH2O 16.9 mg/L 100 uM, Molybdic Acid, Sodium Salt, 2H2O 0.25 mg/L 1.03 uM, Potassium Iodide 0.83 mg/L 5 uM, Potassium, Nitrate 1900 mg/L 18.79 mM, Potassium Phosphate Monobasic 170mg/L 1.25 mM, Zinc Sulfate7H2O 8.6mg/L 29.91 uM, myo-Inositol 100mg/L 0.56 uM, Nicotinic Acid 1 mg/L 8.12 uM, Pyridoxine HCl 1 mg/L 4.86 uM, Thiamine 10mg/L 29.65 uM.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty