Would you like to log in again?
Horse IgG from Horse Serum, MP Biomedicals
Supplier: MP Biomedicals
08641441
IC641441EA 640.82 USD InStock
IC641441
Horse IgG from Horse Serum, MP Biomedicals
Antibodies
Horse IgG is useful for structural and biological studies.
  • Purity greater than 97% by SDS-PAGE
  • Reference standard
  • Starting material for immunogen preparation
  • Supplied purified in Tris or PBS
Immunoglobulin G (IgG) are antibody molecules. Each IgG is composed of four peptide chains - two heavy chains and two light chains. Each IgG has two antigen binding sites. Other Immunoglobulins may be described in terms of polymers with the IgG structure considered the monomer. The fragment antigen-binding (Fab fragment) is a region on an antibody that binds to antigens. It is composed of one constant and one variable domain of each of the heavy and the light chain. These domains shape the paratope — the antigen-binding site — at the amino terminal end of the monomer. The two variable domains bind the epitope on their specific antigens.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty